Lithium chloride, anhydrous 99,9%

Supplier: Apollo Scientific
Warning

IN2323-1KG IN2323-250G
APOSIN2323-1KGEA 759 GBP
APOSIN2323-1KG APOSIN2323-250G
Lithium chloride, anhydrous 99,9%
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2.07 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR