Calcium carbonate 99.999%

Supplier: Apollo Scientific
Warning

Synonyms: Marble

IN1384-25G
APOSIN1384-25GEA 302 GBP
APOSIN1384-25G
Calcium carbonate 99.999%
Calcium carbonate
Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 800 °C (dec.)
Density: 2.93 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR