Anti-IL-2r Mouse Antibody

Supplier: Apollo Scientific
BITP1977-1MG
APOSBITP1977-1MGEA 0 GBP
APOSBITP1977-1MG
Anti-IL-2r Mouse Antibody
Antibodies
Order Now

SPECIFICATIONS

Antigen symbol IL-2r
Antigen name Interleukin-2 receptor
Environmentally Preferable
Reactivity Human
Host Mouse
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR