JC-1 iodide salt 99%

Supplier: Apollo Scientific

Synonyms: 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethylbenzimidazolocarbocyanine iodide, JC-1, 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethyl-imidacarbocyanine iodide, (E)-5,6-Dichloro-2-(3-(5,6-dichloro-1,3-diethyl-1,3-dihydro-2H-benzo[d]imidazol-2-ylidene)prop-1-en-1-yl)-1,3-diethyl-1H-benzo[d]imidazol-3-ium iodide, CBIC2(3), 1,1',3,3'-tetraethyl-5,5',6,6'-tetrachloroimidacarbocyanine iodide

BISN0055-5MG
APOSBISN0055-5MGEA 261 GBP
APOSBISN0055-5MG
JC-1 iodide salt 99%
JC-1 iodide salt

JC-1 is a cyanine dye that has been used to determine mitochondrial transmembrane potential.

Formula: C₂₅H₂₇Cl₄IN₄
MW: 652.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00188475
CAS Number: 47729-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR