G418 disulphate solution 50 mg/ml

Supplier: Apollo Scientific
Warning

Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate

BIG0176-10ML BIG0176-50ML BIG0176-100ML
APOSBIG0176-10MLEA 98.2 GBP
APOSBIG0176-10ML APOSBIG0176-50ML APOSBIG0176-100ML
G418 disulphate solution 50 mg/ml
G418 disulphate
Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR