(S)-2-Acetamido-3-methylbutanoic acid

Supplier: Apollo Scientific
Warning

Synonyms: N-Ac-Val-OH, Ac-Val-OH, N-Acetyl-L-Valine

BIA1796-100G
APOSBIA1796-100GEA 26.2 GBP
APOSBIA1796-100G
(S)-2-Acetamido-3-methylbutanoic acid
(S)-2-Acetamido-3-methylbutanoic acid
Formula: C₇H₁₃NO₃
MW: 159.2 g/mol
Melting Pt: 165 °C
Storage Temperature: Freezer
MDL Number: MFCD00066066
CAS Number: 96-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR