Human Protamine 2 ELISA Kit

Supplier: ANTIBODIES.COM

A313708-96
ANTIA313708-96EA 496 GBP
ANTIA313708-96
Human Protamine 2 ELISA Kit
Assays ELISAs

Human Protamine 2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Protamine 2 in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 1,5 to 24 ng/ml
  • Sensitivity: 0,054 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%


Human Protamine 2 ELISA kit (A313708) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Protamine 2 in serum, plasma or other biological fluids. An antibody specific for Protamine 2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Protamine 2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Protamine 2 antibody, which also binds the Protamine 2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Protamine 2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Protamine 2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Protamine 2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Protamine 2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR