Mouse Circulating Immune Complex ELISA Kit

Supplier: ANTIBODIES.COM

A303560-96
ANTIA303560-96EA 574 GBP
ANTIA303560-96
Mouse Circulating Immune Complex ELISA Kit
Assays ELISAs

Mouse Circulating Immune Complex ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Circulating Immune Complex in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready to use ELISA kit
  • Detection Range: 1,563 to 100 ng/ml
  • Sensitivity: 0,938 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%


Mouse Circulating Immune Complex ELISA kit (A303560) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Circulating Immune Complex in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Circulating Immune Complex has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Circulating Immune Complex present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-Circulating Immune Complex antibody, which binds the captured Circulating Immune Complex present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Circulating Immune Complex captured in each well. The concentration of Circulating Immune Complex can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR