Sleeve protectors, Safe-Knit®
Supplier: Ansell
59-416
ANSE59-416EA 38.1 GBP
ANSE59-416
Sleeve protectors, Safe-Knit®
Workwear
This welder sleeve is made of Kevlar® fabric – combined with an internal cotton layer. It provides protection from the high heat of flying molten particles and sparks.
  • Lightweight and breathable
  • Inner layer made of fire-retardant cotton
  • With thumb holes
Ideal for use in general assembly applications near welding stations and for metal handling in work environments with hot objects.
Certifications: EN 388 (1332), EN 407 (41312X); Cat. III
Order Now
SPECIFICATIONS
Garment application Welding
Environmentally Preferable
Risk area Heat
Gender
Fabric Kevlar® fabric/fire-retardant cotton