Standard hydrometers
Supplier: Alla France
3100MC08020BEO 3500LC10020MEU 3500LC08520BEU 3100MC08020BEU 3100MC09020BEU 3100MC10020MEU 3100MC13020HEU 3100MC14020HEU 3100MC16020HEU 3100MC11020MEU 3100MC12020MEU 3100MC13020MEU 3500MC07020BEU 3500MC07520BEU 3500MC08020BEU 3500MC08520BEU 3500MC09020BEU 3500MC14520BEU 3500MC06520BEU 3500MC14520HEU 3100MC06020BEU 3100MC07020BEU
ALLA3100MC08020BEOEA 1050 GBP
ALLA3100MC08020BEO ALLA3500LC10020MEU ALLA3500LC08520BEU ALLA3100MC08020BEU ALLA3100MC09020BEU ALLA3100MC10020MEU ALLA3100MC13020HEU ALLA3100MC14020HEU ALLA3100MC16020HEU ALLA3100MC11020MEU ALLA3100MC12020MEU ALLA3100MC13020MEU ALLA3500MC07020BEU ALLA3500MC07520BEU ALLA3500MC08020BEU ALLA3500MC08520BEU ALLA3500MC09020BEU ALLA3500MC14520BEU ALLA3500MC06520BEU ALLA3500MC14520HEU ALLA3100MC06020BEU ALLA3100MC07020BEU
Standard hydrometers
Hydrometers
These standard hydrometers strictly adhere to the principles of manufacture and specified calibration.
  • Reading below meniscus
  • Calibration temperature of 20 °C
  • Reference mark on the stem shows any movement of the measurement scale
Certifications: NF, ISO, BS and DIN.
Delivery information: Supplied with certificate of conformity to ISO 649 (corresponding to BS718, DIN12791, NF B 35511).