Femto Pulse 12-Capillary Array
Supplier: AGILENT
A1600-12502240
AGLSA1600-12502240EA 2470 GBP
AGLSA1600-12502240
Femto Pulse 12-Capillary Array
Electrophoresis Systems Capillary Electrophoresis Systems
12-Capillary Array for analysis on the Femto Pulse system.
  • High resolution - optimised length for enhanced resolution; Run Multiple Samples - Run up to 12 samples at same time
  • Low sensitivity - integral part of the Femto Pulse system aiding in low sensitivity detection
The Femto Pulse 12-Capillary Array is used with the Genomic DNA 165 kb kit, the Ultra Sensitivity NGS kit (1 to 6000 bp), the Ultra Sensitivity RNA kit, and the 55 kb BAC kit. It has a separation or effective length of 22 cm, with an overall length of 40 cm. The 12 parallel capillaryMore Product Information