Brilliant III Probe MM, Agilent Technologies
Supplier: AGILENT
600880 600881 600884 600885
732-3798EA 361 GBP
732-3798 AGLS600881 AGLS600884 AGLS600885
Brilliant III Probe MM, Agilent Technologies
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Brilliant III QPCR and QRT-PCR master mixes increase overall sensitivity and speed through the novel, quick-activating hot start and the engineered Taq-mutant.
  • Extremely fast while maintaining sensitivity
  • Greater resistance to common QPCR inhibitors (whole blood or NaCl)
  • Optimized fast cycling formulation ensures reliable and reproducible data with shorter run times
  • Convenient pre-blended formulations compatible with any sequence-specific probe detection chemistry
Brilliant III Ultra-Fast maintains amplification efficiency, R2, dynamic range and detection sensitivity all while shortening the total cycling time by approximately 60% compared to standard cycling.
Caution: For research use only. Not for use in diagnostic procedures.