SideStep II QRT-PCR master mix
Supplier: AGILENT
400917
AGLS400917EA 0 GBP
AGLS400917
SideStep II QRT-PCR master mix
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Complete kit for -step QRT-PCR from cells. Sufficient for 400 reactions.Use Brilliant QRT-PCR technology.
  • Deliver efficient cell lysis
  • RNA stabilization upto 6 months
  • Sensitive quantitative gene expression analysis without RNA purification
  • Purify the most relevant, interesting RNA samples
  • Accurate data: RNA loss minimised
  • Safe: No hazardous waste disposal and removal
These products are ideal for screening multiple samples first, saving more than 70% of the cost and 60% of the time it takes to column purify RNA.
Caution: For research use only. Not for use in diagnostic procedures.