SURE competent cells
Supplier: AGILENT
200152 200227 200238
AGLS200152EA 256 GBP
AGLS200152 AGLS200227 AGLS200238
SURE competent cells
Competent Cells
SURE 2 competent cells are ideal for routine cloning of DNA with secondary structures.
  • Lack components of the pathways that catalyse the rearrangement and deletion of nonstandard secondary and tertiary structures
  • Allows for blue-white colour screening
Replicating eukaryotic DNA in prokaryotic cells can be problematic. Particular eukaryotic genes may contain inverted repeats or secondary structures, such as Z-DNA, that can be rearranged or deleted by E. coli DNA repair systems.
SURE competent cells are deficient in the E. coli genes involved in theMore Product Information