Silver Standard Solution, 1000 mg/L Ag in dil. Nitric Acid Standard for ICP-MS
Supplier: ACTIVE MOTIF
M201.2NP.L1
ACSDM201.2NP.L1EA 145 GBP
ACSDM201.2NP.L1
Silver Standard Solution, 1000 mg/L Ag in dil. Nitric Acid Standard for ICP-MS
Standards Cationic Standards Silver Standards
Ag 1000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Certifications: ISO 17034, ISO 17025, ISO 9001.