Xylenol orange tetrasodium salt ACS

Supplier: Thermo Fisher Scientific

Synonyms: Xylenol orange water soluble, 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)

447600010 447600050 447600250
ACRO447600010EA 77.2 GBP
ACRO447600010 ACRO447600050 ACRO447600250
Xylenol orange tetrasodium salt ACS
Xylenol orange tetrasodium salt
Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7

Order Now

Specification Test Results

Appearance Red-brown to dark brown Crystalline powder
Suitability (for Zn titration) Passes test
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR