Hydrochloric acid 36%, technical

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

444390250
ACRO444390250EA 213 GBP
ACRO444390250
Hydrochloric acid 36%, technical
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 57 °C (760 mmHg)
Melting Pt: –35 °C
Density: 1.16 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Color scale ≤20 APHA
Titration with NaOH 35 to 37 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR