E/E 70%, technical (Not denatured)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethyl alcohol

444280050
ACRO444280050EA 199 GBP
ACRO444280050
E/E 70%, technical (Not denatured)
Ethanol
Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 85…90 °C (760 mmHg)
Melting Pt: –48 °C
Flash Pt: 21 °C°C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3 II
Merck Index: 12,03806

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Density 0.877 to 0.902 kg/l
E/E 67 to 73 % v/v (calculated)
GC ≥99.7 % (ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR