Tetraethylammonium hydroxide 10% aqueous solution

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Tetrylammonium hydroxide

420281000 420285000
ACRO420281000EA 53.1 GBP
ACRO420281000 ACRO420285000
Tetraethylammonium hydroxide 10% aqueous solution
Tetraethylammonium hydroxide
Formula: C₈H₂₁NO
MW: 147.26 g/mol
Density: 0.865
MDL Number: MFCD00009024
CAS Number: 77-98-5
EINECS: 201-073-3
UN: 2922
ADR: 8 II
Merck Index: 12,09342

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl 9.5 to 10.5 %
Melting point 153 to 157 °C
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR