Methyl trifluoromethanesulphonate 96%

Supplier: Thermo Fisher Scientific
Danger

415011000
ACRO415011000EA 594 GBP
ACRO415011000
Methyl trifluoromethanesulphonate 96%
Methyl trifluoromethanesulphonate
Formula: C₂H₃F₃O₃S
MW: 164.11 g/mol
Boiling Pt: 95 °C (760 mmHg)
Density: 1.45 g/cm³
Flash Pt: 38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000409
CAS Number: 333-27-7
EINECS: 206-371-7
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3250 to 1.3270 (20°C, 589 nm)
GC ≥95 %
Enantiomeric excess ≥99.0 % (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR