1-Hexanesulphonic acid sodium salt monohydrate 98% HPLC

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Sodium 1-hexanesulphonate monohydrate

411600050 411600250 411601000 411605000
ACRO411600050EA 144 GBP
ACRO411600050 ACRO411600250 ACRO411601000 ACRO411605000
1-Hexanesulphonic acid sodium salt monohydrate 98% HPLC
1-Hexanesulphonic acid sodium salt monohydrate
Formula: C₆H₁₃NaO₃S·1H₂O
MW: 206.24 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149549
CAS Number: 207300-91-2
EINECS: 220-601-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 7 to 9 %
Solubility (10 % in water) Clear colorless
UV at 250 nm A: ≤0.05
Titration after Ion exchange ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR