Dodecyltrimethylammonium chloride 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC

409320250 409321000
ACRO409320250EA 171 GBP
ACRO409320250 ACRO409321000
Dodecyltrimethylammonium chloride 98%
Dodecyltrimethylammonium chloride
Formula: C₁₅H₃₄ClN
MW: 263.89 g/mol
Melting Pt: 237 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00041974
CAS Number: 112-00-5
EINECS: 203-927-0
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to light yellow Powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Solubility (10 % in water) Clear
Color of solution ≤100 APHA (10 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR