Cerium(IV) sulphate 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate

404340250 404341000
ACRO404340250EA 69.7 GBP
ACRO404340250 ACRO404341000
Cerium(IV) sulphate 99%
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
MW: 332.24 g/mol
Melting Pt: 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
UN: 3260
ADR: 8 II
Merck Index: 12,02036

Order Now

Specification Test Results

Appearance Yellow Powder and/or chunks
Titration Iodometric 39.6 to 44.7 % (Ce)
Refractive index 1.5500 to 1.5540 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR