Buffer solutions, ready to use

Supplier: Thermo Fisher Scientific

383835000 383830010 383855000 383850010 383875000 383870010
ACRO383835000EA 60.2 GBP
ACRO383835000 ACRO383830010 ACRO383855000 ACRO383850010 ACRO383875000 ACRO383870010
Buffer solutions, ready to use
Buffers pH Calibration Buffers

  • Traceable to NIST
  • Clear colourless solutions

Order Now

SPECIFICATIONS

Environmentally Preferable
Density 1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR