Ethynylferrocene 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-cyclopenta-2,4-dienyl-(2-ethynyl-1-cyclopenta-2,4-dienyl)iron, Ferrocene, ethynyl-, (2α)-, (2α)-Ethynylferrocene

378830010
ACRO378830010EA 229 GBP
ACRO378830010
Ethynylferrocene 97%
Ethynylferrocene

Appearance: Red-brown Crystalline powder or crystals

Formula: C₁₂H₁₀Fe
MW: 210.057 g/mol
Melting Pt: 50…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03701501
CAS Number: 1271-47-2

Order Now

Specification Test Results

Appearance Red-brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 50 to 57 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR