2-Iodoxybenzoic acid Stabilised

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Hydroxy-1,2-benziodoxy-3-(1h)-one-1-xoide, IBX, 2-Iodylbenzoic acid

374650100 374650500
ACRO374650100EA 364 GBP
ACRO374650100 ACRO374650500
2-Iodoxybenzoic acid Stabilised
2-Iodoxybenzoic acid

Appearance: White to off-white Powder or prills and/or agglomerates

Formula: C₇H₅IO₄
MW: 280.02 g/mol
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD02912492
CAS Number: 61717-82-6
UN: 1759

Order Now

Specification Test Results

Appearance White to beige Powder or crystals
Titration Iodometric 39 to 50 % as 2-Iodoxybenzoic acid
Identification HPLC (2-Iodoxybenzoic acid, 2-Iodosobenzoic acid, Isophthalic acid, Benzoic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR