tert-Butylperoxy isopropyl carbonate 75% 75% aromatic free mineral spirit

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-[(isopropoxycarbonyl)peroxy]-2-methylpropane, 2-methyl-2-{[(propan-2-yloxy)carbonyl]peroxy}propane, Carbonoperoxoic acid,OO-(1,1-dimethylethyl) O-(1-methylethyl) ester

349842500
ACRO349842500EA 148 GBP
ACRO349842500
tert-Butylperoxy isopropyl carbonate 75% 75% aromatic free mineral spirit
tert-Butylperoxy isopropyl carbonate
Formula: C₈H₁₆O₄
MW: 176.212 g/mol
Melting Pt: –20 °C
Density: 0.905 g/cm³
Flash Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00126878
CAS Number: 2372-21-6
UN: 3103
ADR: 5.2

Order Now

Specification Test Results

Appearance Clear solution
Titration Iodometric 74 to 76 %
Color scale ≤10 APHA
Hydrolyzable chloride ≤150 ppm Inorganic + organic
Impurity ≤1000 ppm Hydroperoxides (as TBHP)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR