Calcium chloride, anhydrous 96%, powder, extra_pure

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

349610250 349615000 349610025 349610100
ACRO349610250EA 34.6 GBP
ACRO349610250 ACRO349615000 ACRO349610025 ACRO349610100
Calcium chloride, anhydrous 96%, powder, extra_pure
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: >1600 °C (760 mmHg)
Melting Pt: 782 °C
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥93.0 %
Sulfate (SO4) ≤500 ppm
Barium (Ba) ≤200 ppm
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤1000 ppm
Heavy metals (as Pb) ≤50 ppm
Insoluble matter ≤0.1 % (in water)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR