n-Hexane 95%, ECD tested halocarbons free grade Residual

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

326710010 326710025
ACRO326710010EA 93.8 GBP
ACRO326710010 ACRO326710025
n-Hexane 95%, ECD tested halocarbons free grade Residual
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –95 °C
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.005 % (Coulometric)
GC ≥94 %
UV at 200 nm A: ≤0.602
Color scale ≤5 APHA
Residue after evaporation ≤0.0002 %
Acidity (CH3COOH) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR