Barium bromide, anhydrous 99%, pure

Supplier: Thermo Fisher Scientific
Warning

315380500 315382500
ACRO315380500EA 132 GBP
ACRO315380500 ACRO315382500
Barium bromide, anhydrous 99%, pure
Barium bromide
Formula: BaBr₂
MW: 297.14 g/mol
Boiling Pt: 1835 °C (1013 hPa)
Melting Pt: 850 °C
Storage Temperature: Ambient
MDL Number: MFCD00003444
CAS Number: 10553-31-8
EINECS: 234-140-0
UN: 1564
ADR: 6.1 III
Merck Index: 12,00995

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR