Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate 98%, pure

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethylenediaminetetraaceticacidferricsodiumsalttrihydrate, (Ethylenedinitrilo)tetraacetic acid ferric sodium salt, EDTA ferric sodium salt

304680010 304682500
ACRO304680010EA 621 GBP
ACRO304680010 ACRO304682500
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate 98%, pure
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate
Formula: C₁₀H₁₂FeN₂NaO₈·3H₂O
MW: 421.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00798117
CAS Number: 18154-32-0
EINECS: 239-802-2

Order Now

Specification Test Results

Appearance Yellow-brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR