1,1,1,3,3,3-Hexafluoro-2-propanol 99.9% Spectroscopy

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

293410100 293410500
ACRO293410100EA 327 GBP
ACRO293410100 ACRO293410500
1,1,1,3,3,3-Hexafluoro-2-propanol 99.9% Spectroscopy
1,1,1,3,3,3-Hexafluoro-2-propanol
Formula: C₃H₂F₆O
MW: 168.04 g/mol
Boiling Pt: 59 °C (760 mmHg)
Melting Pt: –4 °C
Density: 1.596 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 3265
ADR: 8 I
Merck Index: 13,09598

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.01 %
GC ≥99.85 %
UV at 240 nm A: ≤0.009
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR