Ethyl-3-ethoxypropionate 99+% Stabilised, pure

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethyl 3-ethoxypropanoate

269490010 269490025 269490100
ACRO269490010EA 84.9 GBP
ACRO269490010 ACRO269490025 ACRO269490100
Ethyl-3-ethoxypropionate 99+% Stabilised, pure
Ethyl 3-ethoxypropionate
Formula: C₇H₁₄O₃
MW: 146.19 g/mol
Boiling Pt: 170 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.949 g/cm³
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00051356
CAS Number: 763-69-9
EINECS: 212-112-9
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4080 (20°C, 589 nm)
Water ≤0.05 %
GC ≥99.0 %
Color scale ≤15 APHA
Stabilizer 50 to 100 ppm BHT


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR