4-Epioxytetracycline, 97%, can be used as secondary standard

Supplier: Thermo Fisher Scientific

257711000
ACRO257711000EA 424 GBP
ACRO257711000
4-Epioxytetracycline, 97%, can be used as secondary standard
Standards Organic Standards

CAS No.: 14206-58-7


MDL No.: MFCD00021181
GHS Symbols: GHS07, GHS08,
Melting Point: 168 °C (D)
Appearance: Green-beige to brown Powder
Storage Temperature: Ambient
Signal word: Warning

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR