Brij® L23 MBAL-PA-(MV)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Polyoxyethylene (23) Lauryl Ether, Laureth-23, Brij 35

228340010 228342500 228345000 228340051
ACRO228340010EA 189 GBP
ACRO228340010 ACRO228342500 ACRO228345000 ACRO228340051
Brij® L23 MBAL-PA-(MV)
Brij® L23

Previous name Brij® 35

Formula: (C₂₀H₄₂O₅)n
Boiling Pt: >100 °C (760 mmHg)
Melting Pt: 33…40 °C
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00043063
CAS Number: 9002-92-0
EINECS: 500-002-6

Order Now

Specification Test Results

Appearance White Waxy pellets
Infrared spectrum Conforms
Water ≤1 %
Antioxidant without antioxidant
Impurity ≤1 ppm free Ethyleneoxide
Acid value ≤5 mg KOH/g
Hydroxyl value 40 to 60 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR