Sodium chlorate 99+%, extra_pure

Supplier: Thermo Fisher Scientific
Danger

223220010 223222500 223220051
ACRO223220010EA 123 GBP
ACRO223220010 ACRO223222500 ACRO223220051
Sodium chlorate 99+%, extra_pure
Sodium chlorate
Formula: ClNaO₃
MW: 106.44 g/mol
Boiling Pt: 300
Melting Pt: 248…261 °C
Density: 2.49 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1 II
REACH: 01-2119474389-23
Merck Index: 13,08670

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Water ≤0.05 %
Titration ≥99.0 %
Sodium chloride (NaCl) ≤1000 ppm
Insoluble matter ≤0.005 % (in water)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR