Potassium acetate, anhydrous 99+%, pure

Supplier: Thermo Fisher Scientific

Synonyms: Acetic acid potassium salt

220150010 220151000 220150050 220150250
ACRO220150010EA 105 GBP
ACRO220150010 ACRO220151000 ACRO220150050 ACRO220150250
Potassium acetate, anhydrous 99+%, pure
Potassium acetate
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 292 °C
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline or granular powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR