4-(Dimethylamino)phenethyl alcohol 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-[4-(dimethylamino)phenyl]ethanol, 2-(4-(Dimethylamino)phenyl)ethanol, Benzeneethanol, 4-(dimethylamino)-, p-(Dimethylamino)phenethyl alcohol

210930010 210930050
ACRO210930010EA 116 GBP
ACRO210930010 ACRO210930050
4-(Dimethylamino)phenethyl alcohol 99%
4-(Dimethylamino)phenethyl alcohol
Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Boiling Pt: 116 °C (.1 mmHg)
Melting Pt: 53.9…55.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00009711
CAS Number: 50438-75-0

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 58 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR