Tetraphenylphosphonium chloride 98%

Supplier: Thermo Fisher Scientific
Warning

204030100 204030500
ACRO204030100EA 108 GBP
ACRO204030100 ACRO204030500
Tetraphenylphosphonium chloride 98%
Tetraphenylphosphonium chloride
Formula: C₂₄H₂₀ClP
MW: 374.85 g/mol
Melting Pt: 275…280 °C
MDL Number: MFCD00011916
CAS Number: 2001-45-8
EINECS: 217-890-3

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 275 to 280 °C
Titration Argentometric ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR