Sulphur 99.5+%, refined

Supplier: Thermo Fisher Scientific
Warning

201220025 201220100 201225000
ACRO201220025EA 125 GBP
ACRO201220025 ACRO201220100 ACRO201225000
Sulphur 99.5+%, refined
Sulphur
Formula: S
MW: 32.07 g/mol
Boiling Pt: 445 °C (760 mmHg)
Melting Pt: 114 °C
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1 III
Merck Index: 13,09059

Order Now

Specification Test Results

Appearance Yellow Powder
Assay ≥99.50 %
Water ≤0.5 %
Sulfated ash ≤0.08 %
Acidity ≤0.01 % (as H2SO4)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR