Palladium 10% on activated carbon, dry hydrogenation catalyst, unreduced

Supplier: Thermo Fisher Scientific
Warning

195030100 195030500 195030010 195032500
ACRO195030100EA 573 GBP
ACRO195030100 ACRO195030500 ACRO195030010 ACRO195032500
Palladium 10% on activated carbon, dry hydrogenation catalyst, unreduced
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3178
ADR: 4.1 III

Order Now

Specification Test Results

Appearance Black Powder
Assay 9.7 to 10.3 % (Pd)
Loss on drying ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR