Titanium(IV) isopropoxide 98+%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Titanium tetraisopropanolate, Titanium (IV) isopropoxide

194700050 194702500 194700010 194700025
ACRO194700050EA 49.1 GBP
ACRO194700050 ACRO194702500 ACRO194700010 ACRO194700025
Titanium(IV) isopropoxide 98+%
Titanium (IV) isopropoxide
Formula: C₁₂H₂₈O₄Ti
MW: 284.23 g/mol
Boiling Pt: 240 °C (760 mmHg)
Melting Pt: >15 °C
Density: 0.95 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00008871
CAS Number: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3 III
Merck Index: 12,09614

Order Now

Specification Test Results

Appearance Clear liquid (May darken on storage)
Infrared spectrum Conforms
Melting point ≥15 °C
Assay 16.6 to 17.3 % (Ti)
Color scale ≤100 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR