Ethyl cellulose, 10 cps 48% ethoxyl 10 cps

Supplier: Thermo Fisher Scientific

Synonyms: Ethylcellulose

190962500 190965000
ACRO190962500EA 165 GBP
ACRO190962500 ACRO190965000
Ethyl cellulose, 10 cps 48% ethoxyl 10 cps
Ethyl cellulose

Appearance: White to light yellow Crystalline powder or granules

Storage Temperature: Ambient
MDL Number: MFCD00131037
CAS Number: 9004-57-3

Order Now

Specification Test Results

Appearance White to yellow Powder or granules
Infrared spectrum Conforms
Assay 47.5 to 49.5 % (Ethoxyl content)
Viscosity 8 to 12 cP (5% in 80/20 toluene/ethanol, 25°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR