Sodium deuteroxide 30% w/w D2O NMR (D₂O 99+ atom%D)

Supplier: Thermo Fisher Scientific
Danger

172890200
ACRO172890200EA 116 GBP
ACRO172890200
Sodium deuteroxide 30% w/w D2O NMR (D₂O 99+ atom%D)
Sodium deuteroxide
Formula: NaOD
MW: 41.01 g/mol
Density: 1.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00037669
CAS Number: 14014-06-3
EINECS: 237-825-2
UN: 1824
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless Solution
D-Enrichment ≥99.0 %
Concentration 29 to 32 % w/w
Specific optical rotation -2° to -8° (20°C, 589 nm) c=0.5 in Methanol

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR