1-(Trimethylsilyl)imidazole 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: TSIM, N-(Trimethylsilyl)imidazole, N-(Trimethylsilyl)-1H-imidazole, TMSI, 1-(Trimethylsilyl)-1H-imidazole

167130250 167131000 167132500
ACRO167130250EA 104 GBP
ACRO167130250 ACRO167131000 ACRO167132500
1-(Trimethylsilyl)imidazole 97%
1-(Trimethylsilyl)imidazole
Formula: C₆H₁₂N₂Si
MW: 140.26 g/mol
Boiling Pt: 93…94 °C (13 mmHg)
Melting Pt: –42 °C
Density: 0.95 g/cm³
Flash Pt: 5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005280
CAS Number: 18156-74-6
EINECS: 242-040-3
UN: 2733
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4780 (20°C, 589 nm)
GC ≥96.0 %
Insoluble matter ≤0.01 % (10 % in water)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR