(R)-(-)-2-Octanol 99+%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (R)-(-)-2-Hydroxy octane, (R)-2-Octanol, L(-)-2-Octanol, (R)-(-)-Hexyl methyl carbinol, (R)-(-)-Methyl hexyl carbinol

164590050 164590100 164590010
ACRO164590050EA 163 GBP
ACRO164590050 ACRO164590100 ACRO164590010
(R)-(-)-2-Octanol 99+%
(R)-(-)-2-Octanol
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 175 °C (760 mmHg)
Density: 0.83 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00064284
CAS Number: 5978-70-1
EINECS: 227-777-0
UN: 1986
ADR: 3 III
Merck Index: 12,06850

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4270 (20°C, 589 nm)
GC ≥99.0 %
Specific optical rotation -9° to -10° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR