4-Nitrophenol 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Hydroxy-4-nitrobenzene, p-Nitrophenol

157050010 157050050 157051000 157052500 157050051
ACRO157050010EA 185 GBP
ACRO157050010 ACRO157050050 ACRO157051000 ACRO157052500 ACRO157050051
4-Nitrophenol 99%
4-Nitrophenol
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 279 °C (760 mmHg)
Melting Pt: >112 °C
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00007331
CAS Number: 100-02-7
EINECS: 202-811-7
UN: 1663
ADR: 6.1 III
Merck Index: 13,06654

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 112 to 115 °C
GC ≥98.5 %
Titration with NaOH ≥96.0 to ≤104.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR