Benzyl chloroformate 97% w/w Stabilised

Supplier: Thermo Fisher Scientific
Danger

Synonyms: CBZ-Cl (Z-Cl)

152940050 152941000 152940025 152945000
ACRO152940050EA 68.2 GBP
ACRO152940050 ACRO152941000 ACRO152940025 ACRO152945000
Benzyl chloroformate 97% w/w Stabilised
Benzyl chloroformate
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 178…180 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1.2 g/cm³
Flash Pt: 92 °C
Storage Temperature: Freezer
MDL Number: MFCD00000640
CAS Number: 501-53-1
EINECS: 207-925-0
UN: 1739
ADR: 8 I

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5180 (20°C, 589 nm)
Additional info benzyl chloride during storage
GC ≥96.0 %
Color scale ≤100 APHA
Stabilizer 0.1 % anhydrous sodium carbonate
Phosgene ≤0.1 %
Iron (Fe) ≤2 ppm
Acidity (HCl) ≤0.3 %
Benzyl alcohol ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR