1-(2-Pyridylazo)-2-naphthol (PAN) 98%, pure indicator grade

Supplier: Thermo Fisher Scientific
Warning

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

146310050 146310100 146310500
ACRO146310050EA 182 GBP
ACRO146310050 ACRO146310100 ACRO146310500
1-(2-Pyridylazo)-2-naphthol (PAN) 98%, pure indicator grade
1-(2-Pyridylazo)-2-naphthol
Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Boiling Pt: 412
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
EINECS: 201-637-9

Order Now

Specification Test Results

Appearance Orange to red-brown to brown Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
Suitability (as complexom. indicator) Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR