Mineral oil (petroleum) IR, nujol mull preparations_suitable

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Petrolatum, liquid, Mineral oil, Paraffin oil, Nujol

124020010 124020025 124020060 124020100
ACRO124020010EA 105 GBP
ACRO124020010 ACRO124020025 ACRO124020060 ACRO124020100
Mineral oil (petroleum) IR, nujol mull preparations_suitable
Mineral oil (petroleum)
Density: 0.851 g/cm³
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5

Order Now

Specification Test Results

Appearance Clear colorless Oily liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4730 (20°C, 589 nm)
Density (15°C) 830 to 865 kg/l
Viscosity 13 to 19 mm²/s (40°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR